Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_531633.5 | XP_531633.2 | 731 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris |