Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BGRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627773.2 | XP_005627830.1 | 4 - 5 | 856 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |