Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014112008.2 | XP_013967483.1 | 378 | 96 | ||
XM_022416508.1 | XP_022272216.1 | 492 | 96 | |||
XM_536287.5 | XP_536287.3 | 492 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |