Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIH1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619775.3 | XP_005619832.1 | 478 | 97 | ||
XM_022418309.1 | XP_022274017.1 | 684 | 97 | |||
XM_022418312.1 | XP_022274020.1 | 478 | 97 | |||
XM_536575.6 | XP_536575.1 | 458 | 97 |