Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639568.3 | XP_003639616.1 | 4 - 5 | 608 | 101 | |
XM_022404169.1 | XP_022259877.1 | 4 - 5 | 611 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |