Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_539144.4 | XP_539144.2 | 571 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |