Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMTNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022405409.1 | XP_022261117.1 | 3 - 4 | 1058 | 68 | |
XM_022405410.1 | XP_022261118.1 | 2 - 3 | 923 | 68 | ||
XM_540610.6 | XP_540610.2 | 2 - 3 | 887 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |