Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GADD45G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_541314.6 | XP_541314.3 | 1 - 2 | 166 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |