Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014109381.2 | XP_013964856.2 | 2 - 3 | 171 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |