Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639910.3 | XP_005639967.1 | 6 - 7 | 2704 | 105 | |
XM_005639911.3 | XP_005639968.1 | 6 - 7 | 2410 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |