Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022410553.1 | XP_022266261.1 | 4 - 5 | 569 | 116 | |
XM_543564.5 | XP_543564.2 | 4 - 5 | 473 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |