Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCNKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544547.5 | XP_544547.2 | 12 - 13 | 1257 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |