Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPNS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419421.1 | XP_022275129.1 | 12 - 13 | 1366 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |