Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048125.1 | NP_001041590.1 | 1 - 2 | 93 | 83 | |
XM_005641023.3 | XP_005641080.1 | 1 - 2 | 240 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |