Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022421339.1 | XP_022277047.1 | 3 - 4 | 464 | 90 | |
XM_022421342.1 | XP_022277050.1 | 3 - 4 | 224 | 90 | ||
XM_849989.5 | XP_855082.1 | 4 - 5 | 526 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |