Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_850113.5 | XP_855206.1 | 3 - 4 | 1064 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |