Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_540955.6 | XP_540955.2 | 1388 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |