Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_854531.5 | XP_859624.1 | 3 - 4 | 646 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |