Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624278.2 | XP_005624335.1 | 341 | 137 | ||
XM_005624279.2 | XP_005624336.1 | 334 | 137 | |||
XM_014116434.2 | XP_013971909.1 | 261 | 137 | |||
XM_537577.6 | XP_537577.2 | 342 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris |