Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636119.3 | XP_005636176.1 | 563 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |