Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131056.1 | NP_001124528.1 | 881 | 109 | ||
XM_005629474.2 | XP_005629531.1 | 860 | 109 | |||
XM_005629475.3 | XP_005629532.1 | 627 | 109 | |||
XM_005629477.3 | XP_005629534.1 | 1204 | 109 | |||
XM_005629478.3 | XP_005629535.1 | 472 | 109 | |||
XM_014119776.2 | XP_013975251.1 | 635 | 109 | |||
XM_014119777.2 | XP_013975252.1 | 666 | 109 | |||
XM_022403592.1 | XP_022259300.1 | 1363 | 109 | |||
XM_022403593.1 | XP_022259301.1 | 908 | 109 | |||
XM_022403594.1 | XP_022259302.1 | 1272 | 109 | |||
XM_022403595.1 | XP_022259303.1 | 934 | 109 | |||
XM_022403596.1 | XP_022259304.1 | 659 | 109 | |||
XM_022403597.1 | XP_022259305.1 | 537 | 109 | |||
XM_022403598.1 | XP_022259306.1 | 761 | 109 | |||
XM_022403599.1 | XP_022259307.1 | 606 | 109 | |||
XM_022403600.1 | XP_022259308.1 | 529 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |