Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR14L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636489.3 | XP_005636546.1 | 10 | 6582 | 130 | |
XM_014107906.2 | XP_013963381.1 | 9 | 6055 | 130 | ||
XM_014107908.2 | XP_013963383.1 | 8 | 5932 | 130 | ||
XM_022410718.1 | XP_022266426.1 | 10 | 6515 | 130 | ||
XM_022410719.1 | XP_022266427.1 | 10 | 6497 | 130 | ||
XM_022410720.1 | XP_022266428.1 | 9 | 6399 | 130 | ||
XM_022410721.1 | XP_022266429.1 | 9 | 6381 | 130 | ||
XM_534740.6 | XP_534740.2 | 9 | 6460 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |