Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003068.3 | NP_001003068.1 | 2 - 3 | 269 | 68 | |
GenBank mRNA | U65376.2 | - | 2 - 3 | 252 | 68 | |
Z69597.1 | - | 2 - 3 | 145 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |