Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636918.2 | XP_005636975.1 | 2 - 3 | 251 | 154 | |
XM_005636919.3 | XP_005636976.1 | 4 - 5 | 391 | 154 | ||
XM_005636920.2 | XP_005636977.1 | 3 - 4 | 331 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |