Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003263.1 | NP_001003263.1 | 609 | 136 | ||
GenBank mRNA | Z12168.1 | - | 609 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |