Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_541048.5 | XP_541048.2 | 1421 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Canis familiaris |