Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_848160.4 | XP_853253.1 | 1 - 2 | 699 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |