Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS6ST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_847525.5 | XP_852618.1 | 3 - 4 | 1340 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |