Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424829.1 | XP_022280537.1 | 2 - 3 | 216 | 190 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |