Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS2R41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080723.1 | NP_001074192.1 | 890 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris |