Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082421.1 | NP_001075890.1 | 1065 | 61 | ||
GenBank mRNA | EF382361.1 | - | 1065 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris |