Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097551.1 | NP_001091020.1 | 8 - 9 | 668 | 93 | |
XM_022426505.1 | XP_022282213.1 | 8 - 9 | 673 | 93 | ||
GenBank mRNA | EF121758.1 | - | 8 - 9 | 668 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |