Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109960.1 | NP_001103430.1 | 6 - 7 | 450 | 105 | |
XM_014119221.2 | XP_013974696.1 | 6 - 7 | 490 | 105 | ||
GenBank mRNA | AB299408.1 | - | 6 - 7 | 450 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |