Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JMJD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204235.1 | NP_001191164.1 | 3 - 4 | 466 | 86 | |
XM_022412476.1 | XP_022268184.1 | 3 - 4 | 868 | 86 | ||
XM_022412477.1 | XP_022268185.1 | 3 - 4 | 486 | 86 | ||
XM_022412478.1 | XP_022268186.1 | 3 - 4 | 831 | 86 | ||
XM_022412479.1 | XP_022268187.1 | 3 - 4 | 816 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |