Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR30E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049254.1 | 1 | 36 | 64 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-30c-1 | MIR30C-1 | cfa-mir-30c-1 | -3.1kb | Sense | Chr. 15 - 2280050 - 2280110 on NCBI Build CanFam3.1 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |