Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6pd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246727.1 | NP_001233656.1 | 8 - 9 | 994 | 74 | |
GenBank mRNA | AF044676.1 | - | 8 - 9 | 994 | 74 | |
JF935143.1 | - | 7 - 8 | 859 | 74 | ||
JI880984.1 | - | 3 - 4 | 374 | 74 | ||
JP058834.1 | - | 3 - 4 | 374 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |