Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246729.1 | NP_001233658.1 | 1 | 1171 | 91 | |
GenBank mRNA | JI867321.1 | - | 1 | 1201 | 91 | |
JP049411.1 | - | 1 | 1202 | 91 | ||
M34561.1 | - | 1 | 1171 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Both primers and probe map within a single exon |