Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504622.2 | XP_003504670.2 | 4 - 5 | 1893 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |