Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cstf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501440.2 | XP_003501488.1 | 3 - 4 | 556 | 68 | |
XM_007642098.1 | XP_007640288.1 | 3 - 4 | 612 | 68 | ||
GenBank mRNA | JI863144.1 | - | 3 - 4 | 545 | 68 | |
JP056671.1 | - | 3 - 4 | 545 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |