Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511957.2 | XP_003512005.1 | 4 - 5 | 887 | 92 | |
GenBank mRNA | JI883556.1 | - | 4 - 5 | 868 | 92 | |
JP043203.1 | - | 4 - 5 | 867 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |