Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497379.2 | XP_003497427.1 | 1 | 1222 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |