Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508827.2 | XP_003508875.1 | 1 | 526 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |