Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jakmip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513798.2 | XP_003513846.1 | 17 - 18 | 2226 | 75 |