Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103158792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007607948.1 | XP_007606138.1 | 112 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 |