Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654063.1 | XP_007652253.1 | 9 - 10 | 1713 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |