Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamtor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502922.2 | XP_003502970.1 | 3 - 4 | 417 | 60 | |
GenBank mRNA | JI867134.1 | - | 3 - 4 | 359 | 60 | |
JP056109.1 | - | 3 - 4 | 359 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |