Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galtl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652246.1 | XP_007650436.1 | 10 - 11 | 912 | 113 | |
GenBank mRNA | JP038488.1 | - | 10 - 11 | 1012 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |