Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504064.1 | XP_003504112.1 | 14 - 15 | 1710 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |