Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643369.1 | XP_007641559.1 | 3 - 4 | 374 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |