Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515728.2 | XP_003515776.2 | 638 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay |